Please login to your online account to display your discounted pricing

Lithium bis(trimethylsilyl)amide, 95%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  338140100

 View more versions of this product

Catalog No. AC338140100

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Crystalline Powder or Chunks
Formula Weight 167.33
Quantity 10g
Color Light yellow to yellow
Density -3g/mL
Fieser 04,296; 05,393; 07,197; 12,280; 13,165; 14,194
Infrared Spectrum Authentic
Chemical Name or Material Lithium bis(trimethylsilyl)amide
Packaging Glass bottle
Molecular Formula C6H18LiNSi2
CAS 4039-32-1
Assay Percent Range 95%
Linear Formula [(CH3)3Si]2NLi
MDL Number MFCD00008261
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Flammable solid.
Causes severe skin burns and eye damage.
Reacts violently with water.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Lithium bis(trimethylsilyl)amide
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C6H18LiNSi2
Linear Formula [(CH3)3Si]2NLi
Formula Weight 167.33
MDL Number MFCD00008261
Accessories

Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable solid.
Causes severe skin burns and eye damage.
Reacts violently with water.

SDS & Safety Documents