Please login to your online account to display your discounted pricing

Lithium bis(trimethylsilyl)amide, 1M solution in THF/Ethylbenzene, AcroSeal™, ACROS Organics™


Quantity 800mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  347708000

 View more versions of this product

Catalog No. AC347708000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Turbid Solution
Formula Weight 167.33
Quantity 800mL
Color Yellow to brown
Packaging AcroSeal™ Glass Bottle
EINECS Number 223-725-6
TSCA TSCA
Assay 18 to 22% active base (as LiNSi)
Density 0.8900g/mL
Fieser 04,296; 05,393; 07,197; 12,280; 13,165; 14,194
Molecular Formula C6H18LiNSi2
MDL Number MFCD00008261
CAS 4039-32-1
Synonym 1,1,1,3,3,3-Hexamethyldisilazane lithium salt
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage. May cause respiratory irritation. Highly flammable liquid and vapour. May form explosive peroxides.
CAS 109-99-9
Health Hazard 3 GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. Do not breathe dust/fume/gas/mist/vapors/spray. Immediately
CAS 100-41-4
Chemical Name or Material Lithium bis(trimethylsilyl)amide
Name Note 1M Solution in THF/Ethylbenzene

Specifications

Chemical Name or Material Lithium bis(trimethylsilyl)amide
Name Note 1M Solution in THF/Ethylbenzene
CAS 4039-32-1
CAS 109-99-9
CAS 100-41-4
Assay 18 to 22% active base (as LiNSi)
Molecular Formula C6H18LiNSi2
Formula Weight 167.33
MDL Number MFCD00008261
Synonym 1,1,1,3,3,3-Hexamethyldisilazane lithium salt
Accessories

Danger

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. Do not breathe dust/fume/gas/mist/vapors/spray. Immediately

GHS P Statement Causes severe skin burns and eye damage. May cause respiratory irritation. Highly flammable liquid and vapour. May form explosive peroxides.

SDS & Safety Documents