Please login to your online account to display your discounted pricing

Lithium bis(trimethylsilyl)amide, 15% in 2-MeTHF/Ethylbenzene, AcroSeal™, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Lithium bis(trimethylsilyl)amide
CAS 4039-32-1
CAS 96-47-9
CAS 100-41-4
Assay Percent Range 15%
Molecular Formula C6H18LiNSi2
Linear Formula [(CH3)3Si]2NLi
Formula Weight 167.33
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC446111000

 
thermo fisher scientific
AC446111000
Glass Bottle 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC446118000

 
thermo fisher scientific
AC446118000
Glass Bottle 800mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 167.33
Quantity 100mL
Packaging Glass Bottle
Density 0.8560g/mL
Molecular Formula C6H18LiNSi2
CAS 96-47-9
CAS 100-41-4
Chemical Name or Material Lithium bis(trimethylsilyl)amide
Assay Percent Range 15%
Linear Formula [(CH3)3Si]2NLi
CAS 4039-32-1
Health Hazard 1 Danger
Safety and Handling

Danger