missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium bis(trimethylsilyl)amide, 15% in 2-MeTHF/Ethylbenzene, AcroSeal™, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC446118000

 View more versions of this product

Catalog No. AC446118000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Lithium bis(trimethylsilyl)amide
15%
[(CH3)3Si]2NLi
Solubility in water: decomposes
[Li+].C[Si](C)(C)[N-][Si](C)(C)C
167.33
167.33
AcroSeal™ Glass bottle
0.856
−11°C
4039-32-1,96-47-9,100-41-4
C6H18LiNSi2
hexamethyldisilazane lithium salt, lhmds, lihmds, lithium bis trimethylsilyl amide, lithium bis trimethylsilyl azanide, lithium bis-trimethylsilyl amide, lithium hexamethyldisilazane, lithium hexamethyldisilazide, lithiumbis trimethylsilyl amide, unii-rc4n1i108m
YNESATAKKCNGOF-UHFFFAOYSA-N
lithium;bis(trimethylsilyl)azanide
2733832
0.8560g/mL
Solubility in water: decomposes.
800mL

Chemical Identifiers

4039-32-1
167.33
hexamethyldisilazane lithium salt, lhmds, lihmds, lithium bis trimethylsilyl amide, lithium bis trimethylsilyl azanide, lithium bis-trimethylsilyl amide, lithium hexamethyldisilazane, lithium hexamethyldisilazide, lithiumbis trimethylsilyl amide, unii-rc4n1i108m
lithium;bis(trimethylsilyl)azanide
C6H18LiNSi2
YNESATAKKCNGOF-UHFFFAOYSA-N
2733832
[Li+].C[Si](C)(C)[N-][Si](C)(C)C
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
Reacts violently with water.
May form explosive peroxides

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or ha

GHS Signal Word: Danger

EINECSNumber : 223-725-6

Documents