Please login to your online account to display your discounted pricing

Lithium bis(trimethylsilyl)amide, 1.0M sol. in methyl tert-butyl ether, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle in Tin Can

Manufacturer: thermo fisher scientific  345671000

 View more versions of this product

Catalog No. AC345671000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Solution
Formula Weight 167.33
Quantity 100mL
Color Clear to slightly turbid yellow to light brown
Packaging AcroSeal™ Glass Bottle in Tin Can
EINECS Number 223-725-6
TSCA TSCA
Density 0.8000g/mL
Assay 21 to 25% active base (as LiNSi)
Fieser 04,296; 05,393; 07,197; 12,280; 13,165; 14,194; 01,1049; 02,377; 03,262; 04,443; 05,597; 06,530; 07,329; 08,449; 09,420; 10,357; 11,477; 12,441; 13,21; 14,99; 15,290; 16,304; 17,189
Chemical Name or Material Lithium bis(trimethylsilyl)amide
Name Note 1.0M Solution in Methyl tert-Butyl Ether
Molecular Formula C6H18LiNSi2
MDL Number MFCD00008261
CAS 4039-32-1
Synonym 1,1,1,3,3,3-Hexamethyldisilazane lithium salt
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Reacts violently with water.
CAS 1634-04-4
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If exposed or you feel unwell: Call a POISON CENTRE or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Lithium bis(trimethylsilyl)amide
Name Note 1.0M Solution in Methyl tert-Butyl Ether
CAS 4039-32-1
CAS 1634-04-4
Assay 21 to 25% active base (as LiNSi)
Molecular Formula C6H18LiNSi2
Formula Weight 167.33
MDL Number MFCD00008261
Synonym 1,1,1,3,3,3-Hexamethyldisilazane lithium salt
Accessories

Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If exposed or you feel unwell: Call a POISON CENTRE or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Reacts violently with water.

SDS & Safety Documents