missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium bis(trimethylsilyl)amide, 0.9M solution in methylcyclohexane, AcroSeal™, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC380238000

 View more versions of this product

Catalog No. AC380238000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Liquid
800mL
AcroSeal™ Glass Bottle
4039-32-1,108-87-2
Lithium bis(trimethylsilyl)amide
167.33
Clear to hazy colorless to light yellow
0.7700g/mL
MFCD00008261
C6H18LiNSi2

Chemical Identifiers

4039-32-1,108-87-2
167.33
Lithium bis(trimethylsilyl)amide
C6H18LiNSi2
MFCD00008261

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Harmful if inhaled.
Toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
Reacts violently with water.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger