Please login to your online account to display your discounted pricing

Lithium bis(trimethylsilyl)amide, 0.9M solution in methylcyclohexane, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle in Tin Can

Manufacturer: thermo fisher scientific  AC380231000

 View more versions of this product

Catalog No. AC380231000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 167.33
Quantity 100mL
Color Clear to hazy colorless to light yellow
Packaging AcroSeal™ Glass Bottle in Tin Can
Density 0.7700g/mL
Chemical Name or Material Lithium bis(trimethylsilyl)amide
Molecular Formula C6H18LiNSi2
MDL Number MFCD00008261
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Harmful if inhaled.
Toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
Reacts violently with water.
CAS 108-87-2
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 4039-32-1

Specifications

Chemical Name or Material Lithium bis(trimethylsilyl)amide
CAS 4039-32-1
CAS 108-87-2
Molecular Formula C6H18LiNSi2
Formula Weight 167.33
MDL Number MFCD00008261
Accessories

Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Harmful if inhaled.
Toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
Reacts violently with water.

SDS & Safety Documents