Please login to your online account to display your discounted pricing

Lithium bis(trimethylsilyl)amide, 0.9M solution in methylcyclohexane, AcroSeal™, ACROS Organics™

$96.40 - $96.40

Chemical Identifiers

Chemical Name or Material Lithium bis(trimethylsilyl)amide
CAS 4039-32-1
CAS 108-87-2
Molecular Formula C6H18LiNSi2
Formula Weight 167.33
MDL Number MFCD00008261
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC380238000

  SDS
thermo fisher scientific
AC380238000
AcroSeal™ Glass Bottle 800mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC380231000

  SDS
thermo fisher scientific
AC380231000
AcroSeal™ Glass Bottle in Tin Can 100mL Each for $96.40
Description & Specifications

Specifications

Physical Form Liquid
Formula Weight 167.33
Quantity 800mL
Color Clear to hazy colorless to light yellow
Packaging AcroSeal™ Glass Bottle
Density 0.7700g/mL
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Harmful if inhaled.
Toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
Reacts violently with water.
CAS 4039-32-1
MDL Number MFCD00008261
CAS 108-87-2
Chemical Name or Material Lithium bis(trimethylsilyl)amide
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C6H18LiNSi2
Safety and Handling

Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Harmful if inhaled.
Toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
Reacts violently with water.