missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium Bis(oxalato)borate, TCI America™

Manufacturer:  TCI America L028725G

 View more versions of this product

Catalog No. L028725G



Specifications

Specifications

Lithium Bis(oxalato)borate
25g
C4BLiO8
Lithium Bis(oxalate)borate, Lithium Bis(ethanedioato)borate
[Li+].[B-]12(OC(=O)C(=O)O1)OC(=O)C(=O)O2
193.786
193.79
≥95.0% (T)
White
244761-29-3
MFCD07776904
NVQAYVUCVASGDK-UHFFFAOYSA-N
lithium;1,4,6,9-tetraoxa-5-boranuidaspiro[4.4]nonane-2,3,7,8-tetrone
23677815
Crystal-Powder at 20°C

Chemical Identifiers

244761-29-3
193.786
NVQAYVUCVASGDK-UHFFFAOYSA-N
23677815
[Li+].[B-]12(OC(=O)C(=O)O1)OC(=O)C(=O)O2
C4BLiO8
MFCD07776904
Lithium Bis(oxalate)borate, Lithium Bis(ethanedioato)borate
lithium;1,4,6,9-tetraoxa-5-boranuidaspiro[4.4]nonane-2,3,7,8-tetrone
Safety and Handling

Safety and Handling

EINECSNumber : (2)-4075

TSCA : Yes

Documents