missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium Bis(oxalato)borate, TCI America™

$156.36 - $156.36

Chemical Identifiers

CAS 244761-29-3
Molecular Formula C4BLiO8
Molecular Weight (g/mol) 193.786
MDL Number MFCD07776904
InChI Key NVQAYVUCVASGDK-UHFFFAOYSA-N
Synonym Lithium Bis(oxalate)borate, Lithium Bis(ethanedioato)borate
PubChem CID 23677815
IUPAC Name lithium;1,4,6,9-tetraoxa-5-boranuidaspiro[4.4]nonane-2,3,7,8-tetrone
SMILES [Li+].[B-]12(OC(=O)C(=O)O1)OC(=O)C(=O)O2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
L028725G View Documents Tci America
L028725G
25g Each for $156.36
 
Specifications

Chemical Identifiers

244761-29-3
193.786
NVQAYVUCVASGDK-UHFFFAOYSA-N
23677815
[Li+].[B-]12(OC(=O)C(=O)O1)OC(=O)C(=O)O2
C4BLiO8
MFCD07776904
Lithium Bis(oxalate)borate, Lithium Bis(ethanedioato)borate
lithium;1,4,6,9-tetraoxa-5-boranuidaspiro[4.4]nonane-2,3,7,8-tetrone

Specifications

White
244761-29-3
MFCD07776904
NVQAYVUCVASGDK-UHFFFAOYSA-N
lithium;1,4,6,9-tetraoxa-5-boranuidaspiro[4.4]nonane-2,3,7,8-tetrone
23677815
Crystal-Powder at 20°C
Lithium Bis(oxalato)borate
25g
C4BLiO8
Lithium Bis(oxalate)borate, Lithium Bis(ethanedioato)borate
[Li+].[B-]12(OC(=O)C(=O)O1)OC(=O)C(=O)O2
193.786
193.79
≥95.0% (T)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-4075

TSCA : Yes