missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium Bis(nonafluorobutanesulfonyl)imide, TCI America™

Manufacturer:  TCI America L03075G

 View more versions of this product

Catalog No. L03075G



Specifications

Specifications

Lithium Bis(nonafluorobutanesulfonyl)imide
5g
C8HF18LiNO4S2
1759
SVSJFMZWEBYTRE-UHFFFAOYSA-N
lithium;1,1,2,2,3,3,4,4,4-nonafluoro-N-(1,1,2,2,3,3,4,4,4-nonafluorobutylsulfonyl)butane-1-sulfonamide
131868707
Crystal-Powder at 20°C
White
119229-99-1
MFCD06200837
Bis(nonafluorobutanesulfonyl)imide Lithium Salt
[Li].C(C(C(F)(F)S(=O)(=O)NS(=O)(=O)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(C(F)(F)F)(F)F
588.13
587.12
≥95.0% (T)

Chemical Identifiers

119229-99-1
588.13
SVSJFMZWEBYTRE-UHFFFAOYSA-N
131868707
[Li].C(C(C(F)(F)S(=O)(=O)NS(=O)(=O)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(C(F)(F)F)(F)F
C8HF18LiNO4S2
MFCD06200837
Bis(nonafluorobutanesulfonyl)imide Lithium Salt
lithium;1,1,2,2,3,3,4,4,4-nonafluoro-N-(1,1,2,2,3,3,4,4,4-nonafluorobutylsulfonyl)butane-1-sulfonamide
Safety and Handling

Safety and Handling

TSCA : No

Documents