Please login to your online account to display your discounted pricing

Lithium azide, pure, 20% solution in water, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  335360250

 View more versions of this product

Catalog No. AC335360250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 48.96
Quantity 25g
Color Clear colorless
Assay 17 to 23%
TSCA TSCA
EINECS Number 243-177-1
Purity Grade Pure
Density 1.0880g/mL
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula LiN3
Health Hazard 2 GHS H Statement:
Fatal if swallowed.
Contact with acids liberates very toxic gas.
Chemical Name or Material Lithium azide
Name Note 20% Solution in Water
CAS 19597-69-4
MDL Number MFCD00041956
Molecular Formula LiN3

Specifications

Chemical Name or Material Lithium azide
Name Note 20% Solution in Water
CAS 19597-69-4
Purity Grade Pure
Assay 17 to 23%
Molecular Formula LiN3
Linear Formula LiN3
Formula Weight 48.96
MDL Number MFCD00041956
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Fatal if swallowed.
Contact with acids liberates very toxic gas.

SDS & Safety Documents