missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium amide, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 199860050

 View more versions of this product

Catalog No. AC199860050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Lithiumamide
430°C
373°C
7782-89-0
H2LiN
MFCD00011093
15, 5578
Solubility in water: vigorous reaction. Other solubilities: forming lioh and nh3,insoluble in anhydr. ether,benzene and toluene
[Li+].[NH2-]
22.95
22.95
95%
Glass bottle
1.17
White to Gray
95%
94% min. (LiNH2)
LiNH2
01,600; 03,178; 05,392; 13,159; 17,311
hsdb 649, linh2, lithamide, lithium amide, lithium amide 25g, lithium amide li nh2, lithium amide, powder, lithium azanide, lithiumamide, unii-7393omu9lk
AFRJJFRNGGLMDW-UHFFFAOYSA-N
lithium;azanide
24532
Fine Powder
1.1700g/mL
5g

Chemical Identifiers

7782-89-0
22.95
AFRJJFRNGGLMDW-UHFFFAOYSA-N
24532
[Li+].[NH2-]
H2LiN
MFCD00011093
hsdb 649, linh2, lithamide, lithium amide, lithium amide 25g, lithium amide li nh2, lithium amide, powder, lithium azanide, lithiumamide, unii-7393omu9lk
lithium;azanide
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
In contact with water releases flammable gas.
Reacts violently with water.
May form explosive peroxides.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 231-968-4

TSCA : TSCA

SDS Documents