missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium aluminum hydride, pellets, 97%

Manufacturer:  Alfa Aesar™ 8964314

 View more versions of this product

Catalog No. AA8964314



Specifications

Specifications

Lithium aluminum hydride
AlH4Li
UN1410
lithium aluminum hydride, lithium aluminium hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride
[H-].[H-].[H-].[H-].[Li+].[Al+3]
37.954
37.95
97%
Odorless
White to Gray
25g
16853-85-3
MFCD00011075
14,349
BJKLPLABXHXMIM-UHFFFAOYSA-N
aluminum;lithium;hydride
21226445
Pellet
0.917g/mL
Air & Moisture Sensitive
>125°C (decomposition)

Chemical Identifiers

16853-85-3
37.954
BJKLPLABXHXMIM-UHFFFAOYSA-N
21226445
[H-].[H-].[H-].[H-].[Li+].[Al+3]
AlH4Li
MFCD00011075
lithium aluminum hydride, lithium aluminium hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride
aluminum;lithium;hydride
Safety and Handling

Safety and Handling

GHS H Statement
H260-H301-H314-H318
In contact with water releases flammable gases which may ignite spontaneously.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P231+P232-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Handle under inert gas.
Protect from moisture.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: LITHIUM ALUMINUM HYDRIDE

RTECSNumber : BD0100000

TSCA : TSCA

SDS Documents