missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium aluminum hydride, 95% min, Alfa Aesar™

Reducing Agent For Pharmaceuticals, Perfumes, and Fine organic Chemicals; Source of Hydrogen

Manufacturer:  Alfa Aesar 1344214

 View more versions of this product

Catalog No. AA1344214



Specifications

Specifications

Lithium aluminum hydride
95% min.
MFCD00011075
lithium aluminum hydride, lithium aluminium hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride
[H-].[H-].[H-].[H-].[Li+].[Al+3]
37.954
37.95
0.917g/cm3 at 20°C
Gray
16853-85-3
AlH4Li
Light gray powder
BJKLPLABXHXMIM-UHFFFAOYSA-N
aluminum;lithium;hydride
21226445
Powder
Odorless
25g

Chemical Identifiers

16853-85-3
37.954
BJKLPLABXHXMIM-UHFFFAOYSA-N
21226445
[H-].[H-].[H-].[H-].[Li+].[Al+3]
AlH4Li
MFCD00011075
lithium aluminum hydride, lithium aluminium hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride
aluminum;lithium;hydride
Safety and Handling

Safety and Handling

H260-H301-H314-H318: In contact with water releases flammable gases which may ignite spontaneously. Toxic if swallowed. Causes severe skin burns and eye damage. Causes serious eye damage.

DOTInformation : DOT Class: 4.3, Packaging Group: I

EINECSNumber : 240-877-9

Documents