Please login to your online account to display your discounted pricing

Lithium aluminium hydride, 95%, tablets (ca. 0.6 g), ACROS Organics™


Quantity 25g

Manufacturer: thermo fisher scientific  304530250

 View more versions of this product

Catalog No. AC304530250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥94% (LiAlH4)
Physical Form Pellets
Formula Weight 37.95
Quantity 25g
Color White to light gray
TSCA TSCA
RTECS Number BD0100000
EINECS Number 240-877-9
Density 0.9
Fieser 01,581; 02,242; 03,176; 04,291; 05,382; 06,325; 07,196; 08,286; 09,274; 10,236; 11,289; 12,272; 13,61; 14,190; 15,184; 16,133; 17,162
Merck Index 15, 344
Synonym LAH
Name Note 95%
MDL Number MFCD00011075
Chemical Name or Material Lithium aluminium hydride
Assay Percent Range 95%
Linear Formula LiAlH4
Molecular Formula AlH4Li
CAS 16853-85-3
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Lithium aluminium hydride
Name Note 95%
CAS 16853-85-3
Assay ≥94% (LiAlH4)
Assay Percent Range 95%
Molecular Formula AlH4Li
Linear Formula LiAlH4
Formula Weight 37.95
MDL Number MFCD00011075
Synonym LAH
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.

SDS & Safety Documents