missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium aluminium hydride, 95%, in resealable tin cans, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 332100100

 View more versions of this product

Catalog No. AC332100100


 

Specifications Description & Specifications

Specifications

≥94% (LiAlH4)
37.95
White to light gray
01,581; 02,242; 03,176; 04,291; 05,382; 06,325; 07,196; 08,286; 09,274; 10,236; 11,289; 12,272; 13,61; 14,190; 15,184; 16,133; 17,162
In Resealable Tin Cans
LiAlH4
AlH4Li
16853-85-3
LAH
Crystalline Powder
10g
0.9
15, 344
MFCD00011075
95%
Lithium aluminium hydride
Plastic bag in resealable tin can

Chemical Identifiers

16853-85-3
AlH4Li
37.95
LAH
≥94% (LiAlH4)
95%
LiAlH4
MFCD00011075
Lithium aluminium hydride, In Resealable Tin Cans

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 240-877-9

RTECSNumber : BD0100000

TSCA : TSCA