Please login to your online account to display your discounted pricing

Lithium aluminium hydride, 4.0M solution in diethyl ether, AcroSeal™, ACROS Organics™


Quantity 800mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC199518000

 View more versions of this product

Catalog No. AC199518000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage. Harmful if swallowed. May cause drowsiness or dizziness. In contact with water releases flammable gases which may ignite spontaneously. Extremely flammable liquid and vapor. May form explosive peroxides. Repeated exposure may cause skin dryness or cracking.
Health Hazard 3 GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Keep away from any possible contact with water, because of violent reaction and possible flash fire. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Do not breathe dust/fume/gas/mist/vapors/spray.
Formula Weight 37.95
Physical Form Liquid
Quantity 800mL
Color Clear colorless
Packaging AcroSeal™ Glass Bottle
Assay 18.0 to 22.0% (as LiAlH4)
Density 0.7100g/mL
Fieser 01,581; 02,242; 03,176; 04,291; 05,382; 06,325; 07,196; 08,286; 09,274; 10,236; 11,289; 12,272; 13,61; 14,190; 15,184; 16,133; 17,162
Merck Index 15, 344
Synonym LAH
Storage Information Product may develop some turbidity or precipitate
Health Hazard 1 Danger
Name Note 4.0M Solution in Diethyl Ether
CAS 16853-85-3
Chemical Name or Material Lithium aluminium hydride
Molecular Formula AlH4Li
MDL Number MFCD00011075
CAS 60-29-7
Linear Formula LiAIH4

Specifications

Chemical Name or Material Lithium aluminium hydride
Name Note 4.0M Solution in Diethyl Ether
CAS 16853-85-3
CAS 60-29-7
Assay 18.0 to 22.0% (as LiAlH4)
Molecular Formula AlH4Li
Linear Formula LiAIH4
Formula Weight 37.95
MDL Number MFCD00011075
Synonym LAH
Accessories

Danger

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Keep away from any possible contact with water, because of violent reaction and possible flash fire. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Do not breathe dust/fume/gas/mist/vapors/spray.

GHS H Statement
Causes severe skin burns and eye damage. Harmful if swallowed. May cause drowsiness or dizziness. In contact with water releases flammable gases which may ignite spontaneously. Extremely flammable liquid and vapor. May form explosive peroxides. Repeated exposure may cause skin dryness or cracking.

SDS & Safety Documents