Please login to your online account to display your discounted pricing

Lithium Aluminium Hydride, 2.4M Solution in THF, AcroSeal™, ACROS Organics™


Quantity 800mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC377328000

 View more versions of this product

Catalog No. AC377328000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Viscous Liquid
Formula Weight 37.95
Quantity 800mL
Color Clear yellow
Packaging AcroSeal™ Glass Bottle
Assay 9.2 to 10.5% (as LiAlH4)
Density 0.9000g/mL
Merck Index 15, 344
UN Number 1411
Chemical Name or Material Lithium aluminium hydride
Name Note 2.4M Solution in THF
Molecular Formula AlH4Li
Concentration 9.5 to 10.5% (as LiAlH4)
Linear Formula LiAlH4
CAS 16853-85-3
Health Hazard 1 Danger
CAS 109-99-9
Health Hazard 3 GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Keep away from any possible contact with water, because of violent reaction and possible flash fire. Wear protective gloves/protective
MDL Number MFCD00011075
Synonym LAH
Health Hazard 2 GHS P Statement In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapor. Causes severe skin burns and eye damage. May cause respiratory irritation. May form exp

Specifications

Chemical Name or Material Lithium aluminium hydride
Name Note 2.4M Solution in THF
CAS 16853-85-3
CAS 109-99-9
Assay 9.2 to 10.5% (as LiAlH4)
Molecular Formula AlH4Li
Linear Formula LiAlH4
Formula Weight 37.95
MDL Number MFCD00011075
Accessories

Danger

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Keep away from any possible contact with water, because of violent reaction and possible flash fire. Wear protective gloves/protective

GHS P Statement In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapor. Causes severe skin burns and eye damage. May cause respiratory irritation. May form exp

SDS & Safety Documents