missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lithium aluminum Hydride, 2.4M Solution in THF, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 377321000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentSpecifications
Lithium Aluminum hydride | |
2.4M Solution in THF | |
AlH4Li | |
MFCD00011075 | |
9.5 to 10.5% (as LiAlH4) | |
15, 344 | |
Solubility in water: vigorous reaction. | |
[Li+].[AlH4-] | |
37.95 | |
37.95 | |
AcroSeal™ Glass bottle | |
100 mL | |
Viscous Liquid |
Yellow | |
16853-85-3,109-99-9 | |
LiAlH4 | |
1411 | |
0.9000g/mL | |
lithium aluminum hydride, lithium aluminum hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride | |
OCZDCIYGECBNKL-UHFFFAOYSA-N | |
aluminum;lithium;hydride | |
21226445 | |
9.2 to 10.5% (as LiAlH4) | |
0.9 | |
−17°C |
RUO – Research Use Only