Learn More
Lithium aluminium di-n-butylamide, 0.16M solution in 1,2-Dimethoxyethane, Thermo Scientific™
Supplier: Thermo Fisher Scientific 449195000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Lithium aluminium di-n-butylamide | |
-54°C | |
C32H72AlLiN4 | |
lithium aluminum di-n-butylamide, lithium aluminum di-n-butylamide, 0.16m soln. in 1,2-dimethoxyethane, lithium tetrakis dibutylamino alumanuide | |
[Li+].CCCCN(CCCC)[Al-](N(CCCC)CCCC)(N(CCCC)CCCC)N(CCCC)CCCC | |
546.87 | |
546.87 | |
0.86 |
15405-86-4 | |
500mL | |
-6°C | |
ISBVOXPIPNIBOM-UHFFFAOYSA-N | |
lithium;tetrakis(dibutylamino)alumanuide | |
57369578 | |
Glass bottle | |
0.8600g/mL |
Chemical Identifiers
15405-86-4 | |
546.87 | |
lithium aluminum di-n-butylamide, lithium aluminum di-n-butylamide, 0.16m soln. in 1,2-dimethoxyethane, lithium tetrakis dibutylamino alumanuide | |
lithium;tetrakis(dibutylamino)alumanuide |
C32H72AlLiN4 | |
ISBVOXPIPNIBOM-UHFFFAOYSA-N | |
57369578 | |
[Li+].CCCCN(CCCC)[Al-](N(CCCC)CCCC)(N(CCCC)CCCC)N(CCCC)CCCC |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
May damage fertility.
May damage the unborn child.
Harmful if inhaled.
May form explosive peroxides.
GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
GHS Signal Word: Danger
RUO – Research Use Only