missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium acetate hydrate, Puratronic™, 99.998% (metals basis)

Manufacturer:  Alfa Aesar™ 1298206

 View more versions of this product

Catalog No. AA1298206



Specifications

Specifications

Lithium acetate hydrate
(metals basis)
MFCD00013057
lithium acetate, lithium ethanate, quilone, acoli, acetic acid, lithium salt, lithium ethanoate, lithium 1+ ion acetate, lioac, unii-h4278o5flv, acetic acid lithium salt
[Li+].CC(=O)[O-]
65.984
CHEBI:63045
Crystalline
Puratronic™
5g
546-89-4
C2H3LiO2
14,5521
XIXADJRWDQXREU-UHFFFAOYSA-M
lithium;acetate
3474584
65.98 (Anhydrous)
99.998%
57.8°C

Chemical Identifiers

546-89-4
65.984
XIXADJRWDQXREU-UHFFFAOYSA-M
3474584
lithium;acetate
C2H3LiO2
MFCD00013057
lithium acetate, lithium ethanate, quilone, acoli, acetic acid, lithium salt, lithium ethanoate, lithium 1+ ion acetate, lioac, unii-h4278o5flv, acetic acid lithium salt
CHEBI:63045
[Li+].CC(=O)[O-]
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Documents