Learn More
Lithium acetate hydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01298214
Description
Lithium acetate hydrate is used in the laboratory as buffer for gel electrophoresis of DNA and RNA. It is also used to permeabilize the cell wall of yeast for use in DNA transformation. It is a metal acetate for proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Lithium acetate hydrate | |
546-89-4 | |
25 g | |
C2H3LiO2 | |
14,5521 | |
Soluble in water. | |
[Li+].CC([O-])=O | |
65.98 | |
CHEBI:63045 | |
99.998% |
57.8°C | |
Crystalline | |
(metals basis) | |
MFCD00013057 | |
lithium acetate, lithium ethanate, quilone, acoli, acetic acid, lithium salt, lithium ethanoate, lithium 1+ ion acetate, lioac, unii-h4278o5flv, acetic acid lithium salt | |
XIXADJRWDQXREU-UHFFFAOYSA-M | |
lithium;acetate | |
3474584 | |
65.98 (Anhydrous) | |
Puratronic™ |
Chemical Identifiers
546-89-4 | |
65.98 | |
XIXADJRWDQXREU-UHFFFAOYSA-M | |
3474584 | |
[Li+].CC([O-])=O |
C2H3LiO2 | |
MFCD00013057 | |
lithium acetate, lithium ethanate, quilone, acoli, acetic acid, lithium salt, lithium ethanoate, lithium 1+ ion acetate, lioac, unii-h4278o5flv, acetic acid lithium salt | |
CHEBI:63045 |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P280i-P305+P351+P338
H319
EINECSNumber : 208-914-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only