missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lithium acetate, anhydrous, 99%, extra pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 268640050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Lithium acetate, anhydrous | |
280.0°C to 285.0°C | |
Authentic | |
C2H3LiO2 | |
MFCD00013057 | |
02,107 | |
lithium acetate, lithium ethanate, quilone, acoli, acetic acid, lithium salt, lithium ethanoate, lithium 1+ ion acetate, lioac, unii-h4278o5flv, acetic acid lithium salt | |
XIXADJRWDQXREU-UHFFFAOYSA-M | |
lithium;acetate | |
3474584 | |
65.99 | |
Extra Pure | |
Crystals or Powder |
546-89-4 | |
White to Yellow | |
99% | |
CH3COOLi | |
5 kg | |
15,5577 | |
Solubility in water: 408g/L (20°C). Other solubilities: 667g/L (40°C),938g/L (50°C),freely soluble in alcohol | |
[Li+].CC([O-])=O | |
65.98 | |
CHEBI:63045 | |
99% | |
0.1% max. (in water) |
Chemical Identifiers
546-89-4 | |
65.98 | |
XIXADJRWDQXREU-UHFFFAOYSA-M | |
3474584 | |
lithium;acetate |
C2H3LiO2 | |
MFCD00013057 | |
lithium acetate, lithium ethanate, quilone, acoli, acetic acid, lithium salt, lithium ethanoate, lithium 1+ ion acetate, lioac, unii-h4278o5flv, acetic acid lithium salt | |
CHEBI:63045 | |
[Li+].CC([O-])=O |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 208-914-3
RUO – Research Use Only