missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium 1,1,2,2,3,3-Hexafluoropropane-1,3-disulfonimide 98.0+%, TCI America™

$193.61 - $626.00

Chemical Identifiers

CAS 189217-62-7
Molecular Formula C3F6LiNO4S2
Molecular Weight (g/mol) 299.086
MDL Number MFCD06200805
InChI Key VKQJSJGVEJXINS-UHFFFAOYSA-N
Synonym 4,4,5,5,6,6-Hexafluoroperhydro-1,3,2-dithiazine 1,1,3,3-Tetraoxide Lithium Salt, 1,1,2,2,3,3-Hexafluoropropane-1,3-disulfonimide Lithium Salt
PubChem CID 44630347
IUPAC Name lithium;4,4,5,5,6,6-hexafluoro-1$l^{6},3$l^{6}-dithia-2-azanidacyclohexane 1,1,3,3-tetraoxide
SMILES [Li+].C1(C(S(=O)(=O)[N-]S(=O)(=O)C1(F)F)(F)F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H10571G View Documents Tci America
H10571G
1g Each for $193.61
 
H10575G View Documents Tci America
H10575G
5g Each for $626.00
 
Specifications

Chemical Identifiers

189217-62-7
299.086
VKQJSJGVEJXINS-UHFFFAOYSA-N
44630347
[Li+].C1(C(S(=O)(=O)[N-]S(=O)(=O)C1(F)F)(F)F)(F)F
C3F6LiNO4S2
MFCD06200805
4,4,5,5,6,6-Hexafluoroperhydro-1,3,2-dithiazine 1,1,3,3-Tetraoxide Lithium Salt, 1,1,2,2,3,3-Hexafluoropropane-1,3-disulfonimide Lithium Salt
lithium;4,4,5,5,6,6-hexafluoro-1$l^{6},3$l^{6}-dithia-2-azanidacyclohexane 1,1,3,3-tetraoxide

Specifications

White
189217-62-7
MFCD06200805
VKQJSJGVEJXINS-UHFFFAOYSA-N
lithium;4,4,5,5,6,6-hexafluoro-1$l^{6},3$l^{6}-dithia-2-azanidacyclohexane 1,1,3,3-tetraoxide
44630347
Crystal-Powder at 20°C
Lithium 1,1,2,2,3,3-Hexafluoropropane-1,3-disulfonimide
1g
C3F6LiNO4S2
4,4,5,5,6,6-Hexafluoroperhydro-1,3,2-dithiazine 1,1,3,3-Tetraoxide Lithium Salt, 1,1,2,2,3,3-Hexafluoropropane-1,3-disulfonimide Lithium Salt
[Li+].C1(C(S(=O)(=O)[N-]S(=O)(=O)C1(F)F)(F)F)(F)F
299.086
299.09
≥98.0% (T)
Safety and Handling

Safety and Handling

TSCA : No