Please login to your online account to display your discounted pricing

Lisuride maleate, Tocris™

DA receptor agonist; antiparkinson's agent

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material Lisuride maleate
Assay Percent Range >99%
Molecular Formula C20H26N4O·C4H4O4
Formula Weight 446.55
Synonym N'-[(8α)-9, 10-Didehydro-6-methylergolin-8-yl]-N, N-diethylurea maleate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

405250

 
r & d systems
RDS405250
50mg Each for $1,050.42

405210

 
r & d systems
RDS405210
10mg Each for $252.39
Description & Specifications

Specifications

Formula Weight 446.55
Quantity 50mg
Molecular Formula C20H26N4O·C4H4O4
Chemical Name or Material Lisuride maleate
Synonym N'-[(8α)-9, 10-Didehydro-6-methylergolin-8-yl]-N, N-diethylurea maleate
Recommended Storage Desiccate at Room Temperature
CAS 19875-60-6
Assay Percent Range >99%
Safety and Handling

Desiccate at Room Temperature