Please login to your online account to display your discounted pricing

Alfa Aesar™ (±)-Lisofylline, 98%

Chemical Identifiers

Chemical Name or Material (±)-Lisofylline
CAS 6493-06-7
Formula Weight 280.33
MDL Number MFCD00871850
Synonym (±)-1-(5-Hydroxyhexyl)-3,7-dimethylxanthine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67181MA

  SDS
alfa aesar™
J67181MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67181MB

  SDS
alfa aesar™
J67181MB
25mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67181MF

  SDS
alfa aesar™
J67181MF
50mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 280.33
MDL Number MFCD00871850
CAS 6493-06-7
Chemical Name or Material (±)-Lisofylline
Assay 98%
Synonym (±)-1-(5-Hydroxyhexyl)-3,7-dimethylxanthine
Physical Form Solid
Quantity 10mg