missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lisinopril Dihydrate, TCI America™

Manufacturer:  TCI America L02201G

 View more versions of this product

Catalog No. L02201G



Specifications

Specifications

Lisinopril Dihydrate
148°C
83915-83-7
MFCD01698825
BAVDEDVBIHTHJQ-UVJOBNTFSA-N
(2S)-1-[(2S)-6-amino-2-[[(1S)-1-carboxy-3-phenylpropyl]amino]hexanoyl]pyrrolidine-2-carboxylic acid;hydrate
22887897
Crystal-Powder at 20°C
White-Yellow
1g
C21H33N3O6
1-[Nalpha-[(S)-1-Carboxy-3-phenylpropyl]-L-lysyl]-L-proline
C1CC(N(C1)C(=O)C(CCCCN)NC(CCC2=CC=CC=C2)C(=O)O)C(=O)O.O
423.51
405.50
≥98.0% (T)

Chemical Identifiers

83915-83-7
423.51
BAVDEDVBIHTHJQ-UVJOBNTFSA-N
22887897
C1CC(N(C1)C(=O)C(CCCCN)NC(CCC2=CC=CC=C2)C(=O)O)C(=O)O.O
C21H33N3O6
MFCD01698825
1-[Nalpha-[(S)-1-Carboxy-3-phenylpropyl]-L-lysyl]-L-proline
(2S)-1-[(2S)-6-amino-2-[[(1S)-1-carboxy-3-phenylpropyl]amino]hexanoyl]pyrrolidine-2-carboxylic acid;hydrate
Safety and Handling

Safety and Handling

RTECSNumber : TW3589990

TSCA : No

Recommended Storage : Refrigerator

Documents