missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Liranaftate, TCI America™

$340.70 - $1,182.53

Chemical Identifiers

CAS 88678-31-3
Molecular Formula C18H20N2O2S
Molecular Weight (g/mol) 328.43
MDL Number MFCD00865576
InChI Key VPHPQNGOVQYUMG-UHFFFAOYSA-N
Synonym O-(5,6,7,8-Tetrahydronaphthalen-2-yl) (6-Methoxypyridin-2-yl)(methyl)carbamothioate, (6-Methoxypyridin-2-yl)(methyl)carbamothioic Acid O-(5,6,7,8-Tetrahydronaphthalen-2-yl) Ester
PubChem CID 3936
IUPAC Name O-(5,6,7,8-tetrahydronaphthalen-2-yl) N-(6-methoxypyridin-2-yl)-N-methylcarbamothioate
SMILES CN(C1=NC(=CC=C1)OC)C(=S)OC2=CC3=C(CCCC3)C=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
L030150MG View Documents TCI America
L030150MG
50mg Each for $340.70
 
L0301250MG View Documents TCI America
L0301250MG
250mg Each for $1,182.53
 
Specifications

Chemical Identifiers

88678-31-3
328.43
VPHPQNGOVQYUMG-UHFFFAOYSA-N
3936
CN(C1=NC(=CC=C1)OC)C(=S)OC2=CC3=C(CCCC3)C=C2
C18H20N2O2S
MFCD00865576
O-(5,6,7,8-Tetrahydronaphthalen-2-yl) (6-Methoxypyridin-2-yl)(methyl)carbamothioate, (6-Methoxypyridin-2-yl)(methyl)carbamothioic Acid O-(5,6,7,8-Tetrahydronaphthalen-2-yl) Ester
O-(5,6,7,8-tetrahydronaphthalen-2-yl) N-(6-methoxypyridin-2-yl)-N-methylcarbamothioate

Specifications

White-Yellow
50mg
C18H20N2O2S
O-(5,6,7,8-Tetrahydronaphthalen-2-yl) (6-Methoxypyridin-2-yl)(methyl)carbamothioate, (6-Methoxypyridin-2-yl)(methyl)carbamothioic Acid O-(5,6,7,8-Tetrahydronaphthalen-2-yl) Ester
CN(C1=NC(=CC=C1)OC)C(=S)OC2=CC3=C(CCCC3)C=C2
328.43
328.43
≥98.0% (HPLC)
99°C
88678-31-3
MFCD00865576
VPHPQNGOVQYUMG-UHFFFAOYSA-N
O-(5,6,7,8-tetrahydronaphthalen-2-yl) N-(6-methoxypyridin-2-yl)-N-methylcarbamothioate
3936
Crystal-Powder at 20°C
Liranaftate
Safety and Handling

Safety and Handling

RTECSNumber : FD3890000