Please login to your online account to display your discounted pricing

Linomide, Tocris

Immunomodulator with antiangiogenic properties

$221.01 - $904.70

Chemical Identifiers

Chemical Name or Material Linomide
Assay Percent Range >99%
Molecular Formula C18H16N2O3
Formula Weight 308.33
Synonym 1, 2-Dihydro-4-hydroxy-N, N-dimethyl-2-oxo-N-phenyl-3-quinolinecarboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

146150

 
r & d systems
RDS146150
50mg Each for $904.70

146110

 
r & d systems
RDS146110
10mg Each for $221.01
Description & Specifications

Specifications

Formula Weight 308.33
Quantity 50mg
Molecular Formula C18H16N2O3
Assay Percent Range >99%
Synonym 1, 2-Dihydro-4-hydroxy-N, N-dimethyl-2-oxo-N-phenyl-3-quinolinecarboxamide
Chemical Name or Material Linomide
CAS 84088-42-6
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature