Please login to your online account to display your discounted pricing

Linoleoyl Chloride (Pract.), ACROS Organics™

Chemical Identifiers

Chemical Name or Material Linoleoyl chloride
Name Note pract.
CAS 7459-33-8
Molecular Formula C18H31ClO
Linear Formula CH3(CH2)4CHCHCH2CHCH(CH2)7COCl
Formula Weight 298.89
MDL Number MFCD00151403
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC413225000

  SDS
thermo fisher scientific
AC413225000
Glass Bottle in Plastic Container 500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid or Oil
Formula Weight 298.89
Quantity 500mg
Color Dark brown
Packaging Glass Bottle in Plastic Container
EINECS Number 231-233-8
TSCA TSCA
Density 0.9300g/mL
Refractive Index 1.4700 to 1.4900 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Linoleoyl chloride
Name Note pract.
Storage Information This product darkens on storage
Molecular Formula C18H31ClO
Linear Formula CH3(CH2)4CHCHCH2CHCH(CH2)7COCl
MDL Number MFCD00151403
CAS 7459-33-8
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Infrared Spectrum Authentic
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.