missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Linoleic Anhydride, TCI America™

$172.64 - $598.91

Chemical Identifiers

CAS 24909-68-0
Molecular Formula C36H62O3
Molecular Weight (g/mol) 542.889
MDL Number MFCD00010456
InChI Key BDTYMGCNULYACO-WVZYQCMWSA-N
Synonym cis,cis-9,12-Octadecadienoic Anhydride
PubChem CID 15594594
IUPAC Name [(9E,12E)-octadeca-9,12-dienoyl] (9E,12E)-octadeca-9,12-dienoate
SMILES CCCCCC=CCC=CCCCCCCCC(=O)OC(=O)CCCCCCCC=CCC=CCCCCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
L01451G View Documents TCI America
L01451G
1g Each for $172.64
 
L01455G View Documents TCI America
L01455G
5g Each for $598.91
 
Specifications

Chemical Identifiers

24909-68-0
542.889
BDTYMGCNULYACO-WVZYQCMWSA-N
15594594
CCCCCC=CCC=CCCCCCCCC(=O)OC(=O)CCCCCCCC=CCC=CCCCCC
C36H62O3
MFCD00010456
cis,cis-9,12-Octadecadienoic Anhydride
[(9E,12E)-octadeca-9,12-dienoyl] (9E,12E)-octadeca-9,12-dienoate

Specifications

Undesignated
24909-68-0
MFCD00010456
BDTYMGCNULYACO-WVZYQCMWSA-N
[(9E,12E)-octadeca-9,12-dienoyl] (9E,12E)-octadeca-9,12-dienoate
15594594
Clear Liquid at 20°C
Linoleic Anhydride
1g
C36H62O3
cis,cis-9,12-Octadecadienoic Anhydride
CCCCCC=CCC=CCCCCCCCC(=O)OC(=O)CCCCCCCC=CCC=CCCCCC
542.889
542.89
≥90.0% (T)
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Refrigerator