missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Linoleic acid, tech. 75%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1526518
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Linoleic acid | |
0.902 | |
>110°C (230°F) | |
Weak, Characteristic | |
1.4699 | |
MFCD00064241 | |
1727101 | |
14,5505 | |
OYHQOLUKZRVURQ-HZJYTTRNSA-N | |
(9Z,12Z)-octadeca-9,12-dienoic acid | |
5280450 | |
280.45 | |
Technical |
60-33-3 | |
230°C (16 mmHg) | |
75% | |
C18H32O2 | |
CH3(CH2)4CH=CHCH2CH=CH(CH2)7CO2H | |
50 g | |
Air sensitive | |
linoleic acid, linolic acid, telfairic acid, cis,cis-linoleic acid, 9z,12z-octadeca-9,12-dienoic acid, linoleate, grape seed oil, cis,cis-9,12-octadecadienoic acid, cis-9,cis-12-octadecadienoic acid, 9z,12z-linoleic acid | |
CCCCCC=CCC=CCCCCCCCC(=O)O | |
280.452 | |
CHEBI:17351 | |
75% | |
Liquid |
Chemical Identifiers
60-33-3 | |
280.452 | |
OYHQOLUKZRVURQ-HZJYTTRNSA-N | |
5280450 | |
(9Z,12Z)-octadeca-9,12-dienoic acid |
C18H32O2 | |
MFCD00064241 | |
linoleic acid, linolic acid, telfairic acid, cis,cis-linoleic acid, 9z,12z-octadeca-9,12-dienoic acid, linoleate, grape seed oil, cis,cis-9,12-octadecadienoic acid, cis-9,cis-12-octadecadienoic acid, 9z,12z-linoleic acid | |
CHEBI:17351 | |
CCCCCC=CCC=CCCCCCCCC(=O)O |
Safety and Handling
EINECSNumber : 200-470-9
RTECSNumber : RF9990000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only