Please login to your online account to display your discounted pricing

Linoleic acid, free acid, MP Biomedicals™

$38.80 - $38.80

Chemical Identifiers

Chemical Name or Material Linoleic acid
Name Note For GC analysis
CAS 60-33-3
Physical Form Liquid
Molecular Formula C18H32O2
Formula Weight 280.44
Synonym cis-9-cis-12-Octadecadienoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN10218780

 
mp biomedicals
0210218780
100mL Each for $38.80
Description & Specifications

Specifications

Formula Weight 280.44
Assay 0.6587
Quantity 100mL
Physical Form Liquid
Density 0.9022 (water = 1)
Chemical Name or Material Linoleic acid
Name Note For GC analysis
CAS 60-33-3
Molecular Formula C18H32O2
Synonym cis-9-cis-12-Octadecadienoic acid