Please login to your online account to display your discounted pricing

Linoleic acid, free acid, MP Biomedicals™


Quantity 100mL

Manufacturer: mp biomedicals  0210218780

 View more versions of this product

Catalog No. ICN10218780

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 280.44
Assay 0.6587
Quantity 100mL
Physical Form Liquid
Density 0.9022 (water = 1)
Chemical Name or Material Linoleic acid
Name Note For GC analysis
CAS 60-33-3
Molecular Formula C18H32O2
Synonym cis-9-cis-12-Octadecadienoic acid

Specifications

Chemical Name or Material Linoleic acid
Name Note For GC analysis
CAS 60-33-3
Physical Form Liquid
Molecular Formula C18H32O2
Formula Weight 280.44
Synonym cis-9-cis-12-Octadecadienoic acid