LinerGOLD GC Liners for Thermo Scientific™ TRACE™ 1300 Series and Agilent™ Instruments
Using a unique, state-of-the-art deactivation process, Thermo Scientific™ LinerGOLD™ GC liners provide enhanced inertness for a wide range of compounds.
$153.70 - $251.70
Specifications
For Use With (Equipment) GC Injection Port
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With Packing Material Quantity Diameter (Metric) Inner Price Quantity  
Catalog Number Mfr. No. For Use With Packing Material Quantity Diameter (Metric) Inner Price Quantity  
03-151-650 Thermo Fisher Scientific
453A2265UI
Split/Splitless Liner Quartz Wool Pack of 5 4mm Pack of 5 Each for $171.00
03-151-653 Thermo Fisher Scientific
453A1295UI
Split/Splitless Straight Liner No Pack of 5 4mm Pack of 5 Each for $153.70
03-151-656 Thermo Fisher Scientific
453A1345UI
Splitless Liner with Single Taper No Pack of 5 4mm Pack of 5 Each for $211.35
03-151-659 Thermo Fisher Scientific
453A1925UI
Splitless Liner with Single Taper Quartz Wool Pack of 5 4mm Pack of 5 Each for $219.04
03-151-662 Thermo Fisher Scientific
453A1355UI
Splitless Liner with Double Taper No Pack of 5 4mm Pack of 5 Each for $251.70
03-151-665 Thermo Fisher Scientific
453A1255UI
Precision Liner Quartz Wool Pack of 5 4mm Pack of 5 Each for $199.83
03-151-668 Thermo Fisher Scientific
453A1335UI
Direct Straight Liner No Pack of 5 1mm Pack of 5 Each for $186.37
Specifications
Description
Consistency in your GC liner is of paramount importance – you expect to see a consistently high level of performance time after time. Thermo Scientific™ LinerGOLD™ GC liners are manufactured and tested to ensure that you have confidence with every liner that it installed onto your GC system.
  • Increased accuracy and sensitivity in analysis –LinerGOLD prevents the loss of sensitive compounds in analysis
  • Lower detection limits – when analyzing active and sensitive compounds
  • Excellent reproducibility – gives confidence in your results from liner to liner – setting the GOLD standard
Description & Specifications
Specifications
GC Injection Port