Learn More
Linear alkylbenzenesulfonic acid, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 030574A1
Description
Dodecyl Benzene Sulfonic Acid can be used in all types of household, industrial and institutional cleaners including laundry, dishwash, carwash, hard surface and general purpose detergents. It can also be utilized as an intermediate in preparation of emulsifiers and wetting and foaming agents. Unneutralized compound is an effective surfactant in acidic products such as liquid bowl, aluminum and dairy cleaners. It is an important raw material and intermediate used in Organic Synthesis, Pharma synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Linear alkylbenzenesulfonic acid | |
C18H30O3S | |
1 kg | |
p-3-dodecyl benzenesulphonic acid, c10-c16 alkylbenzenesulfonic acid, 3-4-sulfophenyl dodecane, benzenesulfonic acid, 4-1-ethyldecyl, benzenesulfonic acid, c10-16-alkyl derivs., dsstox_cid_7923, 4-dodecan-3-yl benzenesulfonic acid, c10-c16 alkyl benzene sulfonic acid | |
QJRVOJKLQNSNDB-UHFFFAOYSA-N | |
4-dodecan-3-ylbenzenesulfonic acid | |
29249 | |
Liquid |
68584-22-5 | |
MFCD00147445 | |
UN2586 | |
It is soluble in water. 300,000mg/L at 20°C . | |
CCCCCCCCCC(CC)C1=CC=C(C=C1)S(=O)(=O)O | |
326.495 | |
97% |
Chemical Identifiers
68584-22-5 | |
326.495 | |
QJRVOJKLQNSNDB-UHFFFAOYSA-N | |
29249 | |
CCCCCCCCCC(CC)C1=CC=C(C=C1)S(=O)(=O)O |
C18H30O3S | |
MFCD00147445 | |
p-3-dodecyl benzenesulphonic acid, c10-c16 alkylbenzenesulfonic acid, 3-4-sulfophenyl dodecane, benzenesulfonic acid, 4-1-ethyldecyl, benzenesulfonic acid, c10-16-alkyl derivs., dsstox_cid_7923, 4-dodecan-3-yl benzenesulfonic acid, c10-c16 alkyl benzene sulfonic acid | |
4-dodecan-3-ylbenzenesulfonic acid |
Safety and Handling
GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ARYL SULFONIC ACIDS, LIQUID
EINECSNumber : 271-528-9
RTECSNumber : DB4290000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only