missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Linear alkylbenzenesulfonic acid, 97%

Manufacturer:  Alfa Aesar™ 30574D5

 View more versions of this product

Catalog No. AA30574D5



Specifications

Specifications

Linear alkylbenzenesulfonic acid
68584-22-5
MFCD00147445
p-3-dodecyl benzenesulphonic acid, c10-c16 alkylbenzenesulfonic acid, 3-4-sulfophenyl dodecane, benzenesulfonic acid, 4-1-ethyldecyl, benzenesulfonic acid, c10-16-alkyl derivs., dsstox_cid_7923, 4-dodecan-3-yl benzenesulfonic acid, c10-c16 alkylbenzenesulfonic acid, c10-c16 alkylbenzenesulfonic acid, c10-c16 alkyl benzene sulfonic acid
CCCCCCCCCC(CC)C1=CC=C(C=C1)S(=O)(=O)O
326.495
Liquid
3 x 1kg
C18H30O3S
UN2586
QJRVOJKLQNSNDB-UHFFFAOYSA-N
4-dodecan-3-ylbenzenesulfonic acid
29249
97%

Chemical Identifiers

68584-22-5
326.495
QJRVOJKLQNSNDB-UHFFFAOYSA-N
29249
CCCCCCCCCC(CC)C1=CC=C(C=C1)S(=O)(=O)O
C18H30O3S
MFCD00147445
p-3-dodecyl benzenesulphonic acid, c10-c16 alkylbenzenesulfonic acid, 3-4-sulfophenyl dodecane, benzenesulfonic acid, 4-1-ethyldecyl, benzenesulfonic acid, c10-16-alkyl derivs., dsstox_cid_7923, 4-dodecan-3-yl benzenesulfonic acid, c10-c16 alkylbenzenesulfonic acid, c10-c16 alkylbenzenesulfonic acid, c10-c16 alkyl benzene sulfonic acid
4-dodecan-3-ylbenzenesulfonic acid
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ARYL SULFONIC ACIDS, LIQUID

RTECSNumber : DB4290000

TSCA : TSCA

SDS Documents