Please login to your online account to display your discounted pricing

Linear Alkylbenzene Sulfonate, Certified, 1000ppm (1mL = 1mg), LabChem™

$52.69 - $52.69

Chemical Identifiers

Chemical Name or Material Linear Alkylbenzene Sulfonate
CAS 7732-18-5
CAS Max % 99.39
CAS 7664-93-9
CAS Max % 0.5
CAS 25155-30-0
CAS Max % 0.11
Purity Grade Certified Grade
Linear Formula C18H29NaO3S
Formula Weight 348.48
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC161507

 
lab chem
LC161507
125mL Each for $52.69
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 348.48
Color Clear, colorless
Synonym LAS Solution
CAS Max % 0.11
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
CAS Max % 0.5
Packaging Poly Bottle
Quantity 125mL
Chemical Name or Material Linear Alkylbenzene Sulfonate
CAS 7732-18-5
CAS 7664-93-9
Density 1g/mL
CAS Max % 99.39
Concentration 1000ppm (1mL = 1mg)
Physical Form Liquid
CAS 25155-30-0
Storage Information Refrigerate
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Identification Passes Test
Specific Gravity 1g/mL
Solubility Soluble in water
Linear Formula C18H29NaO3S
Decomposition Information Sulfur compounds; Carbon monoxide; Carbon dioxide
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.