Please login to your online account to display your discounted pricing

Thermo Scientific™ Lindberg/Blue M™ Vacuum Ovens


Benefit from a digital control, built-in over-temperature protection and a flexible vacuum/purge/release system for a range of applications with these ovens.

Manufacturer: thermo scientific™  VO1824A

 View more versions of this product

 Alert

Ovens are not suitable for use with hazardous vapor. For use with nonflammable, noncorrosive inert gases only.

Catalog No. 10-549-227

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Vacuum Range 1 x 10-2 torr (10 microns)
Warranty 1 year (parts and labor)
Item Description 127.4L; 120V; 1500w; Painted cabinet; One door, Two shelves
Voltage 120V
Shipping Weight (Metric) 147kg
Height (English) Exterior 35 in.
Chamber Material Corrosion-resistant stainless steel
Shelves 2
Length (Metric) Exterior 61cm
Depth (Metric) Interior 61cm
Width (English) Exterior 27 in.
Shipping Weight (English) 325 lb.
Length (English) Exterior 24 in.
Dimensions (D x W x H) Interior 24 x 18 x 18 in. (61 x 46 x 46cm)
Depth (English) Interior 24 in.
Electrical Requirements 120V 50/60Hz
Temperature Uniformity ±3.5°C
Temperature Range (Metric) 6° to 260°C
Hertz 50/60Hz
Description Capacity: 127.4L; 120V; 1500w; Painted cabinet; One door, Two shelves
Width (Metric) Interior 46cm
Dimensions (L x W x H) Exterior 24 x 27 x 35 in. (61 x 69 x 89cm)
Safety Features Overtemperature Safety System
Capacity 4.5 cu.ft., 127.4L
Width (English) Interior 18 in.
Ports Inert gas port, vacuum port and vent port
Height (English) Interior 18 in.
Capacity (English) 4.5 cu.ft.
Temperature Control Digital Microprocessor Control
Wattage 1500w
Height (Metric) Interior 46cm
Display LED
Finish Painted Steel
Temperature Resolution ±1°C
Width (Metric) Exterior 69cm
Height (Metric) Exterior 89cm
Capacity (Metric) 127.4L

Benefit from a digital control, built-in over-temperature protection and a flexible vacuum/purge/release system with Thermo Scientific™ Lindberg/Blue M™ Vacuum Ovens. These ovens work well with a range of applications including drying, curing, outgassing, aging, process control and other applications requiring elevated temperature in reduced atmospheres or vacuum/purge with nonflammable and inert atmospheres.

Single-Setpoint Digital Microprocessor Control

  • Maximum temperature: 260°C (500°F) with nominal uniformity of ±3.5% of setpoint
  • Simultaneous LED display of setpoint and actual temperature with push-button view; resolution to within 1°C
  • Keypad data entry and pull-out control module for easy service
  • Independent overtemperature safety system factory set with reset pushbutton

Vacuum System

  • 1in.(2.54cm) manifold, chamber rear, exterior, connects to pump or in-house vacuum source
  • With inert gas injection valve, fresh air inlet, vacuum exhaust port, controllable vacuum release vent port
  • 1000 microns (1 torr) capacity
  • Vacuum gauge, vacuum, gas and inlet valves located on front

Construction

  • Corrosion-resistant stainless-steel cabinet includes three aluminum shelves with painted or stainless-steel finish
  • Rear-mounted vacuum manifold connects to pump (not included) or in-house vacuum source
  • Low-watt heating element extends oven life
  • Heavy-gauge steel, dual-wall construction with fiberglass insulation for minimum heat loss in walls and door
  • High-temperature silicone gasket is specially molded for a positive seal
  • Magnetic latch for positive door seal (4.5 cu. ft. capacity models include mechanical latches)
  • Viewing windows: 0.5in.(1.3cm) thick in two smaller capacity models or 0.75in.(1.9cm) thick in 4.5 cu. ft. model; permit observation of work in progress without loss of heat
  • Stainless-steel exterior models come with double door—a solid outer door over inner window door

Includes:

Power cord and aluminum shelves (number of shelves based on model; see ordering table)