Please login to your online account to display your discounted pricing
Thermo Scientific™ Lindberg/Blue M™ Multipurpose 1500°C Box Furnaces
Lindberg/Blue M multipurpose furnaces feature integral control to 1500°C. Double-wall insulated construction provides rapid heatup/cooldown and energy efficiency.
Specifications
Hertz 50/60Hz
Temperature Range (Metric) 1500°C
Voltage 208/240V
Electrical Requirements 208/240V 50/60Hz
View More Specs
Products
Catalog Number Mfr. No. Description Capacity (Metric) Temperature Control Price Quantity    
10-549-205
thermo scientific™
BF51433C1
Box Furnace 6L Multi Seg/1 Prog CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
Specifications
Hertz 50/60Hz
Temperature Range (Metric) 1500°C
Voltage 208/240V
Electrical Requirements 208/240V 50/60Hz
Thermo Scientific™ Lindberg/Blue M Multipurpose 1500°C Box Furnaces have double-wall construction with Moldatherm™ insulation for rapid heatup and cooldown, energy efficiency and cooler exterior surface temperatures.
Not available in Europe
  • Built-in adjustable high limit overtemperature protection
  • Simultaneous LED display of actual and setpoint temperatures in either °C or °F
  • Silicon carbide heating elements for long-life, safety and reliable service with maximum energy savings
  • Safety door switch interrupts power to heating elements when door is opened; protects elements and minimizes exposure to operator
  • Moldatherm hearthplate supports load and protects interior from spillage and mishandling
  • Type “R” thermocouple is integrated into chamber backwall
With Integral Controller
  • Microprocessor-based PID control
  • Single program with multiple segments for ramp (up and down) and dwell (timed hold) temperature control
With Independent Controller (CC58125C, available separately)
  • Microprocessor-based PID (proportional, integral, derivative) control
  • Single segment, single setpoint, one ramp to setpoint
Requires:
BF51333C requires CC58125C controller