missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lindberg/Blue M™ Box and Tube Furnace Controllers

Program segments into Thermo Scientific™ Lindberg/Blue M™ Box and Tube Furnace Controllers for increased temperature accuracy.

Supplier:  Thermo Scientific™ CC58114PBA1

 View more versions of this product

Catalog No. 15-453-427

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Program segments into Thermo Scientific™ Lindberg/Blue M™ Box and Tube Furnace Controllers for increased temperature accuracy. Features overtemperature control and multiple segment configuration; designed to work with the 1200°C split-hinge and 1500°C box furnaces.

  • Fully wired with advanced microprocessor-based digital control, solid-state power module, on/off curcuit breaker and thermocouple input jacks for each zone
  • Built-in adjustable high-limit overtemperature protection
  • LED simultaneously displays actual temperature vs. setpoint in °C or °F
  • Designed for operation on 120, 208, or 240V 50/60Hz, single-phase line

Overtemperature Control

  • Adjustable digital control is factory installed
  • Protects furnace and load in the event of primary control curcuit failure
  • Overrides main controller and shuts off power to furnace if high limit is reached
  • Must be manually reset for safety
  • Operates via magnetic contacts through a signal from an independent thermocouple

1200°C Three Zone, 16 Segment, One Programmable Controller for Split-hinge Furnaces (CC58434C)

  • Three microprocessor-based PID controls, single segment, single setpoint, one ramp to set point

1200°C Three Zone, 16 Segment, Three Programmable Controllers for Split-hinge Furnaces (CC58434PC)

  • Center Zone—One microprocessor-based PID programmable control, with one program of 16 segments for ramps (up and down) and eight dwells (timed holds)
  • End Zones—Two microprocessor-based PID control, single segment, single setpoint; microprocessers mimic the programmed profile of the center zone controller, but allow a digitally displayed offset up to 100°C

1200°C Digital, Single Setpoint Controllers for Split-hinge Furnaces (CC58114PBA, CC58114PA)

  • Includes one microprocessor-based PID control, single segment, single setpoint, one ramp to setpoint
  • CC58114PBA includes overtemperature control

1500°C Digital, Single Setpoint Controller for Box Furnaces (CC58125C)

  • Includes three microprocessor-based PID controls, single segment, single setpoint, one ramp to set point

1500°C Digital, Single Program, Multiple Segment Programmable Controllers for Box Furnaces (CC58125PC, CC58125PBC)

  • Advanced microprocessor-based PID control, single program with multiple segments for ramp and dwell temperature control
  • CC58125PBC includes overtemperature control
Specifications

Specifications

Single-zone, Multiple Segment, programmable, over temperature control
1200°C (Split-Hinge), 1500°C (Box Furnaces)
Digital
30 A
Lindberg/Blue M 1200°C Split-Hinge Tube Furnaces
Tube Furnace Controller
120 V
50/60 Hz
SDS
Documents

Documents

Product Certifications
Promotions

Promotions