missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lincomycin Hydrochloride Monohydrate, TCI America™

$270.42 - $904.47

Chemical Identifiers

CAS 7179-49-9
Molecular Formula C18H37ClN2O7S
Molecular Weight (g/mol) 461.011
MDL Number MFCD00167245
InChI Key LFZGYTBWUHCAKF-DCNJEFSFSA-N
Synonym Lincocin Hydrochloride
PubChem CID 60209196
IUPAC Name hydron;(2S,4R)-N-[(1R,2R)-2-hydroxy-1-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide;chloride;hydrate
SMILES [H+].CCCC1CC(N(C1)C)C(=O)NC(C2C(C(C(C(O2)SC)O)O)O)C(C)O.O.[Cl-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
L01665G View Documents TCI America
L01665G
5g Each for $270.42
 
L016625G View Documents TCI America
L016625G
25g Each for $904.47
 
Specifications

Chemical Identifiers

7179-49-9
461.011
LFZGYTBWUHCAKF-DCNJEFSFSA-N
60209196
[H+].CCCC1CC(N(C1)C)C(=O)NC(C2C(C(C(C(O2)SC)O)O)O)C(C)O.O.[Cl-]
C18H37ClN2O7S
MFCD00167245
Lincocin Hydrochloride
hydron;(2S,4R)-N-[(1R,2R)-2-hydroxy-1-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide;chloride;hydrate

Specifications

7179-49-9
MFCD00167245
LFZGYTBWUHCAKF-DCNJEFSFSA-N
hydron;(2S,4R)-N-[(1R,2R)-2-hydroxy-1-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide;chloride;hydrate
60209196
Crystal-Powder at 20°C
White
5g
C18H37ClN2O7S
Lincocin Hydrochloride
[H+].CCCC1CC(N(C1)C)C(=O)NC(C2C(C(C(C(O2)SC)O)O)O)C(C)O.O.[Cl-]
461.011
443.00
≥98.0% (T)
154°C
Lincomycin Hydrochloride Monohydrate
Safety and Handling

Safety and Handling

RTECSNumber : RH6315500

TSCA : No