missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lincomycin hydrochloride, 10mg/mL in distilled water, sterile-filtered, Thermo Scientific™
Supplier: Thermo Scientific Chemicals J67091AC
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsLincomycin hydrochloride is an antibiotic used to study the inhibition of protein synthesis, bacterial antibiotic resistance, and protein and surfactant binding 1,2 It is produced by Streptomyces lincolnensis var. lincolnensis and has been used in the treatment of Staphylococcal, Streptococcal, and Bacteroides fragilisinfections. Lincomycin inhibits bacterial protein synthesis by forming cross-links within the peptidyl transferase loop region of the 23S rRNA
Solubility
Soluble in water.
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Specifications
Lincomycin Hydrochloride | |
10mg/mL in distilled water, sterile-filtered | |
859-18-7 | |
MFCD00058237 | |
POUMFISTNHIPTI-OXVORJCQSA-N | |
(2R,4R)-N-[(1R,2R)-2-hydroxy-1-[(2S,3R,4S,5R,6S)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide;hydrochloride | |
124080936 |
Lincomycin hydrochloride | |
Liquid | |
C18H35ClN2O6S | |
Soluble in water. | |
CCCC1CC(N(C1)C)C(=O)NC(C2C(C(C(C(O2)SC)O)O)O)C(C)O.Cl | |
442.996 | |
25 mL |
Chemical Identifiers
859-18-7 | |
442.996 | |
POUMFISTNHIPTI-OXVORJCQSA-N | |
(2R,4R)-N-[(1R,2R)-2-hydroxy-1-[(2S,3R,4S,5R,6S)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide;hydrochloride |
C18H35ClN2O6S | |
MFCD00058237 | |
124080936 | |
CCCC1CC(N(C1)C)C(=O)NC(C2C(C(C(C(O2)SC)O)O)O)C(C)O.Cl |
Safety and Handling
EINECSNumber : 212-726-7
RTECSNumber : RH6315000
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C