Please login to your online account to display your discounted pricing

Alfa Aesar™ Lincomycin hydrochloride, 10mg/mL in distilled water, sterile-filtered

Chemical Identifiers

Chemical Name or Material Lincomycin hydrochloride
CAS 859-18-7
Name Note 10mg/mL in distilled water, sterile-filtered
MDL Number MFCD00058237
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67091AC

  SDS
alfa aesar™
J67091AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67091AD

  SDS
alfa aesar™
J67091AD
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Lincomycin hydrochloride
CAS 859-18-7
MDL Number MFCD00058237
Physical Form Liquid
Storage Note 1 -30° to -10°C
RTECS Number RH6315000
Quantity 25mL
Name Note 10mg/mL in distilled water, sterile-filtered

Applications
Lincomycin hydrochloride is an antibiotic used to study the inhibition of protein synthesis, bacterial antibiotic resistance, and protein and surfactant binding 1,2 It is produced by Streptomyces lincolnensis var. lincolnensis and has been used in the treatment of Staphylococcal, Streptococcal, and Bacteroides fragilisinfections. Lincomycin inhibits bacterial protein synthesis by forming cross-links within the peptidyl transferase loop region of the 23S rRNA

Solubility
Soluble in water.

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Safety and Handling

-30° to -10°C