Please login to your online account to display your discounted pricing

Alfa Aesar™ Lincomycin hydrochloride, 10mg/mL in distilled water, sterile-filtered


Quantity 50mL

Manufacturer: alfa aesar™  J67091AD

 View more versions of this product

Catalog No. AAJ67091AD

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Lincomycin hydrochloride
Physical Form Liquid
CAS 859-18-7
Name Note 10mg/mL in distilled water, sterile-filtered
MDL Number MFCD00058237
Storage Note 1 -30° to -10°C
Quantity 50mL
RTECS Number RH6315000

Specifications

Chemical Name or Material Lincomycin hydrochloride
CAS 859-18-7
Name Note 10mg/mL in distilled water, sterile-filtered
MDL Number MFCD00058237

Applications
Lincomycin hydrochloride is an antibiotic used to study the inhibition of protein synthesis, bacterial antibiotic resistance, and protein and surfactant binding 1,2 It is produced by Streptomyces lincolnensis var. lincolnensis and has been used in the treatment of Staphylococcal, Streptococcal, and Bacteroides fragilisinfections. Lincomycin inhibits bacterial protein synthesis by forming cross-links within the peptidyl transferase loop region of the 23S rRNA

Solubility
Soluble in water.

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Accessories

-30° to -10°C