missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Linalyl Butyrate, TCI America™

Manufacturer:  TCI America L027525ML

 View more versions of this product

Catalog No. L027525ML



Specifications

Specifications

Linalyl Butyrate
Yellow
78-36-4
MFCD00048716
FHLGUOHLUFIAAA-UHFFFAOYSA-N
3,7-dimethylocta-1,6-dien-3-yl butanoate
62321
Clear Liquid at 20°C
82°C
25mL
C14H24O2
Butyric Acid Linalyl Ester
CCCC(=O)OC(C)(CCC=C(C)C)C=C
224.344
224.34
≥97.0% (GC)

Chemical Identifiers

78-36-4
224.344
FHLGUOHLUFIAAA-UHFFFAOYSA-N
62321
CCCC(=O)OC(C)(CCC=C(C)C)C=C
C14H24O2
MFCD00048716
Butyric Acid Linalyl Ester
3,7-dimethylocta-1,6-dien-3-yl butanoate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0763

TSCA : Yes

Documents