missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Linalyl Butyrate, TCI America™

$42.78 - $158.89

Chemical Identifiers

CAS 78-36-4
Molecular Formula C14H24O2
Molecular Weight (g/mol) 224.344
MDL Number MFCD00048716
InChI Key FHLGUOHLUFIAAA-UHFFFAOYSA-N
Synonym Butyric Acid Linalyl Ester
PubChem CID 62321
IUPAC Name 3,7-dimethylocta-1,6-dien-3-yl butanoate
SMILES CCCC(=O)OC(C)(CCC=C(C)C)C=C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
L02755ML View Documents TCI America
L02755ML
5mL Each for $42.78
 
L027525ML View Documents TCI America
L027525ML
25mL Each for $158.89
 
Specifications

Chemical Identifiers

78-36-4
224.344
FHLGUOHLUFIAAA-UHFFFAOYSA-N
62321
CCCC(=O)OC(C)(CCC=C(C)C)C=C
C14H24O2
MFCD00048716
Butyric Acid Linalyl Ester
3,7-dimethylocta-1,6-dien-3-yl butanoate

Specifications

82°C
5mL
C14H24O2
Butyric Acid Linalyl Ester
CCCC(=O)OC(C)(CCC=C(C)C)C=C
224.344
224.34
≥97.0% (GC)
Yellow
78-36-4
MFCD00048716
FHLGUOHLUFIAAA-UHFFFAOYSA-N
3,7-dimethylocta-1,6-dien-3-yl butanoate
62321
Clear Liquid at 20°C
Linalyl Butyrate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0763

TSCA : Yes