missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(+)-Limonene, TCI America™

Manufacturer:  TCI America L01055ML

 View more versions of this product

Catalog No. L01055ML



Specifications

Specifications

(+)-Limonene
5989-27-5
MFCD00062991
d-limonene, +-limonene, r-+-limonene, +-4r-limonene, +-carvene, +-dipentene, citrene, +-p-mentha-1,8-diene, 4r-limonene, d-+-limonene
CC1=CCC(CC1)C(=C)C
136.238
CHEBI:15382
Clear Liquid at 20°C
≥99.0% (GC)
Colorless
5mL
C10H16
2319
XMGQYMWWDOXHJM-JTQLQIEISA-N
(4R)-1-methyl-4-prop-1-en-2-ylcyclohexene
440917
136.24
176°C
-75°C

Chemical Identifiers

5989-27-5
136.238
XMGQYMWWDOXHJM-JTQLQIEISA-N
440917
(4R)-1-methyl-4-prop-1-en-2-ylcyclohexene
C10H16
MFCD00062991
d-limonene, +-limonene, r-+-limonene, +-4r-limonene, +-carvene, +-dipentene, citrene, +-p-mentha-1,8-diene, 4r-limonene, d-+-limonene
CHEBI:15382
CC1=CCC(CC1)C(=C)C
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2245

RTECSNumber : GW6360000

TSCA : Yes

Documents