Please login to your online account to display your discounted pricing

Lime Water, Saturated, LabChem™

$40.00 - $40.00

Chemical Identifiers

Chemical Name or Material Lime Water
CAS 7732-18-5
CAS Max % 99.8
CAS 1305-62-0
CAS Max % 0.2
Name Note Saturated
Linear Formula Ca(OH)2
Formula Weight 74.09
UN Number UN3266
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC161602

 
lab chem
LC161602
Poly Bottle 1L Each for $40.00
Description & Specifications

Specifications

Formula Weight 74.09
Chemical Name or Material Lime Water
Packaging Poly Bottle
Name Note Saturated
Physical Form Liquid
Linear Formula Ca(OH)2
Color Clear, colorless
CAS 7732-18-5
UN Number UN3266
Quantity 1L
Solubility Information Soluble in water
Health Hazard 1 Warning
CAS Max % 99.8
Health Hazard 2 GHS H Statement
Causes eye irritation.
CAS 1305-62-0
Health Hazard 3 GHS P Statement
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Identification Passes Test
Decomposition Information None
CAS Max % 0.2
Recommended Storage Room Temperature
Safety and Handling

Warning

GHS P Statement
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
Causes eye irritation.

Room Temperature